Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144843.4 | NP_659092.1 | 9, 10 | 1229 | ||
GenBank mRNA | AB517623.1 | - | 10, 11 | 1210 | ||
AK076218.1 | - | 9, 10 | 1206 | |||
AK154859.1 | - | 9, 10 | 1149 | |||
BC020019.1 | - | 9, 10 | 1163 | 3596732 |