Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000293.2 | NP_000284.1 | 13 | 1329 | ||
NM_001031835.2 | NP_001027005.1 | 14 | 1441 | |||
XM_005255983.4 | XP_005256040.1 | 13 | 1292 | |||
XM_005255984.4 | XP_005256041.1 | 14 | 1592 | |||
XM_017023282.1 | XP_016878771.1 | 7 | 586 | |||
XM_017023283.1 | XP_016878772.1 | 14 | 1414 | |||
XM_017023284.1 | XP_016878773.1 | 13 | 1295 | |||
GenBank mRNA | BC033657.1 | - | 14 | 1420 | 3913693 | |
CR936710.1 | - | 14 | 1473 | |||
X84908.1 | - | 13 | 1301 |