Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145465.2 | NP_663440.1 | 2 | 535 | ||
GenBank mRNA | AK154596.1 | - | 2 | 640 | ||
AK154959.1 | - | 2 | 536 | |||
AY188357.1 | - | 2 | 545 | |||
BC004650.1 | - | 2 | 367 | 3482980 |