Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172606.2 | NP_766194.2 | 5, 6 | 618 | ||
GenBank mRNA | AK089827.1 | - | 5, 6 | 620 | ||
AK132461.1 | - | 5, 6 | 590 | |||
BU516606.1 | - | 5, 6 | 634 | |||
CF182201.1 | - | 2, 3 | 106 |