Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172121.1 | NP_742119.1 | 1, 2 | 66 | ||
GenBank mRNA | AJ516034.1 | - | 1, 2 | 66 | ||
AK138281.1 | - | 1, 2 | 74 | |||
AK193148.1 | - | 1, 2 | 29 | |||
AK205610.1 | - | 1, 2 | 29 | |||
AK210610.1 | - | 1, 2 | 29 | |||
BC060682.1 | - | 1, 2 | 65 | 6416246 | ||
BC111448.1 | - | 1, 2 | 66 |