Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145139.2 | NP_660121.2 | 5 | 429 | ||
GenBank mRNA | AA038997.1 | - | 1 | 28 | ||
AJ310346.1 | - | 5 | 439 | |||
AK032936.1 | - | 5 | 417 | |||
AK075584.1 | - | 5 | 435 | |||
AK150895.1 | - | 5 | 424 | |||
AK151175.1 | - | 5 | 424 | |||
AK167640.1 | - | 5 | 424 | |||
AK168009.1 | - | 5 | 427 | |||
AK168295.1 | - | 5 | 426 | |||
BC024463.1 | - | 5 | 429 | 4975621 |