Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145139.2 | NP_660121.2 | 11, 12 | 1620 | ||
GenBank mRNA | AJ310346.1 | - | 11, 12 | 1630 | ||
AK032936.1 | - | 11, 12 | 1608 | |||
AK075584.1 | - | 11, 12 | 1626 | |||
AK150895.1 | - | 11, 12 | 1615 | |||
AK151175.1 | - | 11, 12 | 1615 | |||
AK167640.1 | - | 11, 12 | 1615 | |||
AK168009.1 | - | 11, 12 | 1618 | |||
AK168295.1 | - | 11, 12 | 1617 | |||
BC021351.1 | - | 2288 | ||||
BC024463.1 | - | 11, 12 | 1620 | 4975621 |