Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205353.1 | NP_001192282.1 | 17 | 1824 | ||
NM_172611.4 | NP_766199.1 | 17 | 1726 | |||
GenBank mRNA | AK036832.1 | - | 17 | 1715 | ||
AK129209.1 | - | 13 | 3701 | |||
AK142266.1 | - | 16 | 2475 | |||
BC021523.1 | - | 13 | 1100 | |||
BC085148.1 | - | 17 | 1782 | |||
BC139228.1 | - | 17 | 1715 | 8862250 | ||
BC139229.1 | - | 17 | 1715 | 8862251 |