Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145475.4 | NP_663450.3 | 5 | 1030 | ||
GenBank mRNA | AB079067.1 | - | 5 | 537 | ||
AK042077.1 | - | 5 | 621 | |||
AK052269.1 | - | 5 | 796 | |||
AK129416.1 | - | 2 | 172 | |||
AK169528.1 | - | 5 | 615 | |||
AK188338.1 | - | 3 | 260 | |||
BC094253.1 | - | 5 | 1029 | 6400872 |