Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145475.4 | NP_663450.3 | 12 | 1854 | ||
GenBank mRNA | AB079067.1 | - | 12 | 1361 | ||
AK042077.1 | - | 12 | 1445 | |||
AK052269.1 | - | 12 | 1620 | |||
AK129416.1 | - | 9 | 996 | |||
AK161801.1 | - | 692 | ||||
AK169528.1 | - | 12 | 1439 | |||
AK213964.1 | - | 2 | 220 | |||
BC094253.1 | - | 12 | 1853 | 6400872 |