Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027905.2 | NP_082181.2 | 2 | 709 | ||
GenBank mRNA | AK005048.1 | - | 2 | 709 | ||
BC117817.1 | - | 3 | 224 | |||
BC117818.1 | - | 3 | 257 | |||
BC141293.1 | - | 2 | 778 | 9055905 |