Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083114.1 | NP_001076583.1 | 7 | 751 | ||
NM_001285863.1 | NP_001272792.1 | 7 | 880 | |||
NM_001285864.1 | NP_001272793.1 | 6 | 659 | |||
NM_146062.3 | NP_666174.2 | 9 | 1015 | |||
NM_175363.4 | NP_780572.1 | 8 | 808 | |||
GenBank mRNA | AK081606.1 | - | 8 | 801 | ||
AK150904.1 | - | 8 | 759 | |||
AK158488.1 | - | 7 | 881 | |||
AK160138.1 | - | 8 | 781 | |||
AK168443.1 | - | 7 | 753 | |||
BC031486.1 | - | 9 | 968 | 3157250 | ||
BC150854.2 | - | 6 | 655 | 9087765 | ||
CF734096.1 | - | 6 | 639 |