Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY189815.1 | - | 34 | 5343 | ||
BC121792.1 | - | 34 | 5274 | 40099393 |