Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177810.1 | NP_001171281.1 | 4 | 615 | ||
NM_001177811.1 | NP_001171282.1 | 4 | 615 | |||
NM_144850.2 | NP_659099.2 | 4 | 615 | |||
GenBank mRNA | AK044566.1 | - | 4 | 538 | ||
AK142594.1 | - | 4 | 509 | |||
AK169430.1 | - | 4 | 614 | |||
BC020532.1 | - | 4 | 627 | 4236136 |