Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153416.2 | NP_700465.2 | 15 | 1529 | ||
GenBank mRNA | AK034591.1 | - | 15 | 1424 | ||
AK079849.1 | - | 14 | 1537 | |||
AK083401.1 | - | 15 | 1443 | |||
AK083537.1 | - | 15 | 1450 | |||
AK087134.1 | - | 15 | 1505 | |||
AK088247.1 | - | 15 | 1446 | |||
AK134406.1 | - | 15 | 1420 | |||
AK165243.1 | - | 15 | 1444 | |||
AK166118.1 | - | 15 | 1444 | |||
AK169523.1 | - | 15 | 1481 | |||
BC018191.1 | - | 15 | 1430 | 3994248 | ||
BC025501.1 | - | 15 | 1476 | 5321013 | ||
BC026604.1 | - | 14 | 1983 |