Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146065.1 | NP_666177.1 | 10 | 1060 | ||
GenBank mRNA | AK039648.1 | - | 1052 | |||
AK047901.1 | - | 1036 | ||||
AK154588.1 | - | 1051 | ||||
AK155046.1 | - | 10 | 1051 | |||
AK157628.1 | - | 1090 | ||||
BC026483.1 | - | 10 | 1060 | 4503463 |