Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146065.1 | NP_666177.1 | 10, 11 | 1202 | ||
GenBank mRNA | AK039648.1 | - | 1194 | |||
AK047901.1 | - | 1178 | ||||
AK154588.1 | - | 1193 | ||||
AK155046.1 | - | 10, 11 | 1193 | |||
AK157628.1 | - | 1232 | ||||
BC026483.1 | - | 10, 11 | 1202 | 4503463 |