Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144852.3 | NP_659101.2 | 2 | 374 | ||
GenBank mRNA | AK030586.1 | - | 2 | 378 | ||
AK043772.1 | - | 2 | 375 | |||
AK162398.1 | - | 2 | 418 | |||
BC016100.1 | - | 2 | 468 | 4911158 |