Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144852.3 | NP_659101.2 | 5 | 1932 | ||
GenBank mRNA | AK030586.1 | - | 4 | 2217 | ||
AK043772.1 | - | 5 | 1933 | |||
AK162398.1 | - | 5 | 1976 | |||
BC016100.1 | - | 6 | 1550 | 4911158 |