Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178069.5 | NP_835170.1 | 11 | 1457 | ||
GenBank mRNA | AK030940.1 | - | 11 | 1432 | ||
AK078501.1 | - | 11 | 1430 | |||
AK135345.1 | - | 1452 | ||||
AK145186.1 | - | 11 | 1435 | |||
AK146895.1 | - | 11 | 1432 | |||
AK178616.1 | - | 2 | 46 | |||
BC043724.1 | - | 11 | 1457 | 5004563 | ||
BC080306.1 | - | 9 | 1188 |