Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poglut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300827.1 | NP_001287756.1 | 3 | 594 | ||
NM_172380.4 | NP_759012.1 | 3 | 391 | |||
GenBank mRNA | AK031608.1 | - | 3 | 330 | ||
AK035948.1 | - | 3 | 339 | |||
AK036224.1 | - | 3 | 321 | |||
AK041321.1 | - | 3 | 329 | |||
AK045278.1 | - | 3 | 539 | |||
AK079886.1 | - | 3 | 360 | |||
AK082556.1 | - | 3 | 561 | |||
AK167641.1 | - | 3 | 561 | |||
AK204867.1 | - | 1 | 20 | |||
BC026809.1 | - | 3 | 342 | 4205469 | ||
CB249259.1 | - | 3 | 392 |