Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145482.3 | NP_663457.2 | 7 | 807 | ||
GenBank mRNA | AY037804.1 | - | 7 | 807 | ||
BC029101.1 | - | 7 | 704 |