Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252468.1 | NP_001239397.1 | 17 | 2854 | ||
NM_001252469.1 | NP_001239398.1 | 18 | 2765 | |||
NM_057171.2 | NP_476512.1 | 18 | 2908 | |||
GenBank mRNA | AK148796.1 | - | 17 | 2774 | ||
AK164266.1 | - | 18 | 2738 | |||
AK170134.1 | - | 19 | 2833 | |||
BC026647.1 | - | 10 | 1649 |