Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bag6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252468.1 | NP_001239397.1 | 22 | 3380 | ||
NM_001252469.1 | NP_001239398.1 | 23 | 3291 | |||
NM_057171.2 | NP_476512.1 | 23 | 3434 | |||
GenBank mRNA | AK148796.1 | - | 22 | 3300 | ||
AK164266.1 | - | 23 | 3264 | |||
AK170134.1 | - | 24 | 3359 | |||
AK189151.1 | - | 2 | 99 | |||
BC026647.1 | - | 15 | 2175 | |||
BC030296.1 | - | 4 | 303 |