Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-M10.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177634.1 | NP_808302.1 | 2 | 320 | ||
GenBank mRNA | AF539600.1 | - | 2 | 320 | ||
AY211081.1 | - | 2 | 308 |