Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172621.2 | NP_766209.1 | 4 | 651 | ||
GenBank mRNA | AK017800.1 | - | 4 | 612 | ||
AK040195.1 | - | 3 | 289 | |||
AK046522.1 | - | 4 | 640 | |||
AK156849.1 | - | 4 | 492 | |||
BC064037.1 | - | 4 | 636 | 1227040 |