Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002641.3 | NP_002632.1 | 6 | 1455 | ||
NM_020473.3 | NP_065206.3 | 5 | 678 | |||
NR_033835.1 | 6 | 1081 | ||||
NR_033836.1 | 6 | 797 | ||||
XM_011545539.2 | XP_011543841.1 | 5 | 2279 | |||
XM_017029581.1 | XP_016885070.1 | 5 | 1402 | |||
GenBank mRNA | AK098047.1 | - | 5 | 891 | ||
AK303538.1 | - | 5 | 670 | |||
AK315063.1 | - | 5 | 1414 | |||
BC038236.1 | - | 5 | 1412 | 5733899 | ||
D11466.1 | - | 6 | 1424 | |||
S74936.1 | - | 4 | 897 |
Gene Symbol: PIGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | S78467.1 | - | 1 | 1404 |