Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177374.1 | NP_001170845.1 | 14, 15 | 1982 | ||
NM_146078.3 | NP_666190.2 | 14, 15 | 1982 | |||
GenBank mRNA | AK083320.1 | - | 14, 15 | 2121 | ||
AK122254.2 | - | 13, 14 | 2864 | |||
AK143202.1 | - | 14, 15 | 1982 | |||
AK147445.1 | - | 14, 15 | 1971 | |||
AY061885.1 | - | 14, 15 | 1669 | |||
AY280958.1 | - | 14, 15 | 1697 | |||
BC075642.1 | - | 15, 16 | 1796 | 30544183 |