Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177374.1 | NP_001170845.1 | 38 | 4538 | ||
NM_146078.3 | NP_666190.2 | 38 | 4538 | |||
GenBank mRNA | AK078002.1 | - | 6 | 445 | ||
AK122254.2 | - | 36 | 5367 | |||
AK143202.1 | - | 38 | 4538 | |||
AK147445.1 | - | 38 | 4527 | |||
AY061885.1 | - | 38 | 4225 | |||
AY280958.1 | - | 38 | 4253 | |||
BC025617.1 | - | 3 | 192 | |||
BC026391.1 | - | 12 | 1291 | |||
BC031403.1 | - | 23 | 2332 | 4935220 | ||
BC044903.1 | - | 24 | 2473 | |||
BC075642.1 | - | 39 | 4352 | 30544183 |