Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002642.3 | NP_002633.1 | 2 | 1063 | ||
NM_153747.1 | NP_714969.1 | 2 | 1040 | |||
GenBank mRNA | AK308201.1 | - | 2 | 1020 | ||
BC006539.2 | - | 2 | 1010 | 2823399 | ||
BT006734.1 | - | 1 | 729 | |||
CR450292.1 | - | 1 | 729 | |||
D85418.1 | - | 1 | 1022 |