Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcng3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153512.1 | NP_705732.1 | 2 | 1405 | ||
GenBank mRNA | AF454551.1 | - | 2 | 1372 | ||
AF454552.1 | - | 2 | 1405 |