Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146298.2 | NP_001139770.1 | 7 | 1116 | ||
NM_001282093.1 | NP_001269022.1 | 8 | 1266 | |||
NM_153085.4 | NP_694725.3 | 8 | 1425 | |||
GenBank mRNA | AF320996.1 | - | 8 | 1205 | ||
AK044675.1 | - | 1 | 98 | |||
AK087826.1 | - | 8 | 1182 | |||
AK156780.1 | - | 8 | 1250 | |||
AK172416.1 | - | 8 | 1249 | |||
AK173274.1 | - | 7 | 818 | |||
BC020146.1 | - | 7 | 928 | |||
BC057178.1 | - | 8 | 1057 | |||
BC080851.1 | - | 8 | 1383 | 6848738 | ||
BG083129.2 | - | 1 | 65 | |||
CO432980.1 | - | 2 | 180 |