Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153552.3 | NP_705780.1 | 13 | 1076 | ||
GenBank mRNA | AK031785.1 | - | 13 | 1077 | ||
AK032200.1 | - | 13 | 1077 | |||
AK042867.1 | - | 13 | 1082 | |||
AK047991.1 | - | 9 | 2339 | |||
AK052835.1 | - | 13 | 1072 | |||
AK139307.1 | - | 13 | 1082 | |||
AK167374.1 | - | 13 | 1090 | |||
AK168619.1 | - | 13 | 1341 | |||
BC024951.1 | - | 13 | 1248 | 4010605 | ||
BC027149.1 | - | 7 | 658 |