Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145492.4 | NP_663467.1 | 4 | 3337 | ||
GenBank mRNA | AK052655.1 | - | 4 | 3241 | ||
AK077697.1 | - | 1 | 159 | |||
AK081096.1 | - | 1 | 794 | |||
AK131130.1 | - | 3 | 3422 | |||
AY147406.1 | - | 4 | 3240 | |||
BC021376.1 | - | 4 | 3153 | 5038671 |