Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002644.3 | NP_002635.2 | 9 | 2237 | ||
XM_011509629.1 | XP_011507931.1 | 9 | 2148 | |||
GenBank mRNA | AF272149.2 | - | 9 | 2233 | ||
BC110494.2 | - | 9 | 2134 | 40007650 | ||
BC110495.2 | - | 9 | 2134 | 40007652 | ||
CR749533.1 | - | 10 | 2310 | |||
M24559.1 | - | 7 | 1840 | |||
S62403.1 | - | 9 | 2209 | |||
X73079.1 | - | 9 | 2233 |