Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lims2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144862.3 | NP_659111.1 | 9, 10 | 911 | ||
GenBank mRNA | BC010816.1 | - | 9, 10 | 912 | 4212305 |