Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146086.2 | NP_666198.1 | 21 | 2516 | ||
GenBank mRNA | AK042142.1 | - | 5 | 376 | ||
BC031925.1 | - | 21 | 2516 | 4501123 | ||
BC044892.1 | - | 21 | 2516 | 4527430 | ||
BC158126.1 | - | 21 | 2700 | 9007394 | ||
X60664.1 | - | 21 | 2524 |