Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146086.2 | NP_666198.1 | 13 | 1744 | ||
GenBank mRNA | BC031925.1 | - | 13 | 1744 | 4501123 | |
BC044892.1 | - | 13 | 1744 | 4527430 | ||
BC158126.1 | - | 13 | 1928 | 9007394 | ||
X60664.1 | - | 13 | 1752 |