Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172632.2 | NP_766220.2 | 5 | 1827 | ||
GenBank mRNA | AK079654.1 | - | 5 | 1805 | ||
BC058942.1 | - | 4 | 1230 | 6821544 | ||
BC062911.1 | - | 4 | 1230 | 6821544 | ||
BC094564.1 | - | 4 | 2041 |