Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139269.2 | NP_644675.2 | 4, 5 | 540 | ||
GenBank mRNA | AB298806.1 | - | 2, 3 | 385 | ||
AK039703.1 | - | 4, 5 | 753 | |||
AK042007.1 | - | 4, 5 | 493 | |||
AK163505.1 | - | 4, 5 | 541 | |||
AX775891.1 | - | 4, 5 | 522 | |||
BC024581.1 | - | 4, 5 | 486 | 5065247 |