Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024560.2 | NP_001019731.1 | 5 | 524 | ||
GenBank mRNA | AK037118.1 | - | 515 | |||
AK047234.1 | - | 5 | 499 | |||
AK143441.1 | - | 4 | 636 | |||
BC048922.1 | - | 5 | 524 | 6311864 |