Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001984.2 | NP_001001984.2 | 17 | 3524 | ||
GenBank mRNA | AK141779.1 | - | 3 | 765 | ||
AK173084.1 | - | 6 | 1342 | |||
BC036264.1 | - | 5 | 1223 | |||
BC037117.1 | - | 10 | 2073 | |||
BC057051.1 | - | 4 | 897 | |||
BC076576.1 | - | 17 | 3089 | 6840591 |