Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eml3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144872.1 | NP_659121.1 | 9 | 1282 | ||
GenBank mRNA | BC022146.1 | - | 9 | 1282 | 5101626 | |
BC023878.1 | - | 9 | 1305 | 5354867 | ||
BC034112.1 | - | 9 | 1080 |