Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035239.2 | NP_001030316.1 | 5, 6 | 813 | ||
NM_001035240.2 | NP_001030317.1 | 5, 6 | 813 | |||
NM_001035241.2 | NP_001030318.1 | 5, 6 | 813 | |||
NM_001035242.1 | NP_001030319.1 | 5, 6 | 813 | |||
NM_001035243.2 | NP_001030320.1 | 5, 6 | 813 | |||
NM_001035244.1 | NP_001030321.1 | 5, 6 | 861 | |||
NM_001035245.1 | NP_001030322.1 | 5, 6 | 861 | |||
NM_001035246.1 | NP_001030323.1 | 5, 6 | 861 | |||
NM_177341.4 | NP_796315.2 | 5, 6 | 880 | |||
GenBank mRNA | AJ544532.1 | - | 5, 6 | 813 | ||
AJ544533.1 | - | 5, 6 | 813 | |||
AJ544534.1 | - | 5, 6 | 813 | |||
AJ544535.1 | - | 5, 6 | 813 | |||
AJ544536.1 | - | 5, 6 | 813 | |||
AK044454.1 | - | 5, 6 | 882 | |||
AK044533.1 | - | 5, 6 | 659 | |||
AK046908.1 | - | 5, 6 | 694 | |||
AK051934.1 | - | 5, 6 | 659 | |||
AK052064.1 | - | 5, 6 | 667 | |||
AK053673.1 | - | 5, 6 | 698 | |||
AK077331.1 | - | 5, 6 | 693 | |||
AK082141.1 | - | 5, 6 | 695 | |||
AK084402.1 | - | 5, 6 | 666 | |||
AK142504.1 | - | 5, 6 | 694 | |||
AK161909.1 | - | 5, 6 | 694 | |||
AK173218.1 | - | 5, 6 | 884 | |||
BC037449.1 | - | 5, 6 | 709 | |||
BC141400.1 | - | 5, 6 | 856 | 9056013 | ||
BC141412.1 | - | 5, 6 | 856 | 9056025 | ||
HM064407.1 | - | 5, 6 | 905 | |||
HM064408.1 | - | 5, 6 | 905 | |||
HM064409.1 | - | 5, 6 | 905 | |||
HM064410.1 | - | 5, 6 | 905 | |||
HM064411.1 | - | 5, 6 | 905 | |||
JF706722.1 | - | 5, 6 | 876 | |||
JX644970.1 | - | 5, 6 | 905 | |||
JX644971.1 | - | 5, 6 | 905 | |||
JX644972.1 | - | 5, 6 | 905 | |||
JX644973.1 | - | 5, 6 | 905 | |||
JX644974.1 | - | 5, 6 | 905 | |||
JX644975.1 | - | 5, 6 | 905 | |||
JX644976.1 | - | 5, 6 | 905 | |||
JX644977.1 | - | 5, 6 | 905 | |||
JX644978.1 | - | 5, 6 | 905 | |||
JX644979.1 | - | 5, 6 | 905 | |||
JX644980.1 | - | 5, 6 | 905 | |||
JX644981.1 | - | 5, 6 | 905 | |||
JX644982.1 | - | 5, 6 | 905 | |||
JX644983.1 | - | 5, 6 | 905 | |||
JX644984.1 | - | 5, 6 | 905 | |||
JX644985.1 | - | 5, 6 | 905 | |||
JX644986.1 | - | 5, 6 | 905 |