Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099634.1 | NP_001093104.1 | 42, 43 | 5030 | ||
GenBank mRNA | AK028612.1 | - | 14, 15 | 1671 | ||
AK075903.1 | - | 8, 9 | 813 | |||
AK173131.1 | - | 42, 43 | 4811 | |||
BC025649.1 | - | 16, 17 | 1887 | |||
BC044825.1 | - | 19, 20 | 2553 | |||
BC150790.1 | - | 43, 44 | 4843 | 9087701 |