Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099634.1 | NP_001093104.1 | 35 | 4238 | ||
GenBank mRNA | AK028612.1 | - | 7 | 879 | ||
AK041571.1 | - | 7 | 866 | |||
AK075903.1 | - | 1 | 21 | |||
AK173131.1 | - | 35 | 4019 | |||
BC025649.1 | - | 9 | 1095 | |||
BC044825.1 | - | 18 | 2436 | |||
BC150790.1 | - | 36 | 4051 | 9087701 | ||
CN701681.1 | - | 5 | 580 |