Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145500.3 | NP_663475.1 | 3 | 620 | ||
GenBank mRNA | AK078479.1 | - | 3 | 620 | ||
BC016129.1 | - | 3 | 446 | 4920477 |