Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198108.2 | NP_932776.1 | 3 | 301 | ||
GenBank mRNA | AK154147.1 | - | 3 | 289 | ||
AK159230.1 | - | 3 | 288 | |||
BB871690.1 | - | 3 | 302 | |||
BC023055.1 | - | 3 | 268 | |||
BC057060.1 | - | 2 | 677 | |||
CV563048.1 | - | 3 | 227 |