Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153796.3 | NP_722491.2 | 5 | 2489 | ||
GenBank mRNA | AK054159.1 | - | 5 | 787 | ||
AK146244.1 | - | 5 | 2383 | |||
AY059385.1 | - | 5 | 1951 | |||
BC034909.1 | - | 4 | 811 | |||
BC071195.1 | - | 5 | 2489 | 5100486 |