Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164263.1 | NP_001157735.1 | 4 | 285 | ||
NM_172641.3 | NP_766229.2 | 4 | 285 | |||
GenBank mRNA | AK036908.1 | - | 4 | 283 | ||
AK053921.1 | - | 4 | 283 | |||
BC090250.1 | - | 4 | 263 | 6485933 |